missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRS1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of IRS1.
Brand: Abnova PAB7671.100uL
This item is not returnable.
View return policy
Specifications
| IRS1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| IRS1 | |
| IRS1 | |
| A synthetic peptide corresponding to residues surrounding S639 of human IRS1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of IRS1. | |
| In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| HIRS-1 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 3667 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction