Learn More
ITCH, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human ITCH.
Brand: Abnova PAB29228.100uL
Description
Atrophin-1 contains a polyglutamine repeat, expansion of which is responsible for dentatorubral and pallidoluysian atrophy. The protein encoded by this gene interacts with atrophin-1. This encoded protein is a closely related member of the NEDD4-like protein family. This family of proteins are E3 ubiquitin-ligase molecules and regulate key trafficking decisions, including targeting of proteins to proteosomes or lysosomes. This encoded protein contains four tandem WW domains and a HECT (homologous to the E6-associated protein carboxyl terminus) domain. It can act as a transcriptional corepressor of p45/NFE2 and may participate in the regulation of immune responses by modifying Notch-mediated signaling. It is highly similar to the mouse Itch protein, which has been implicated in the regulation and differentiation of erythroid and lymphoid cells. [provided by RefSeq
Specifications
| ITCH | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human ITCH. | |
| In PBS, pH 7.4 (without Mg2+ and Ca2+), (50% glycerol, 0.02% sodium azide) | |
| AIF4/AIP4/NAPP1/dJ468O1.1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 83737 | |
| Human, Mouse | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| ITCH | |
| ITCH | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y420 of human ITCH. | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of ITCH only when phosphorylated at tyrosine 420. | |
| Store at -20°C. Avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only