Learn More
ITGAX Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ITGAX.
Brand: Abnova PAB16498.50ug
Description
This gene encodes the integrin alpha X chain protein. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles. [provided by RefSeq
Specifications
| ITGAX | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ITGAX. | |
| In PBS (0.09% sodium azide) | |
| CD11C/SLEB6 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 3687 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| ITGAX | |
| ITGAX | |
| A synthetic peptide (conjugated with KLH) corresponding to human ITGAX. | |
| 50 μg | |
| Primary | |
| Internal domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only