Learn More
ITGB1 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ITGB1.
Brand: Abnova PAB16504.50ug
Description
Integrins are heterodimeric proteins made up of alpha and beta subunits. At least 18 alpha and 8 beta subunits have been described in mammals. Integrin family members are membrane receptors involved in cell adhesion and recognition in a variety of processes including embryogenesis, hemostasis, tissue repair, immune response and metastatic diffusion of tumor cells. This gene encodes a beta subunit. Multiple alternatively spliced transcript variants which encode different protein isoforms have been found for this gene. [provided by RefSeq
Specifications
| ITGB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ITGB1. | |
| In PBS (0.09% sodium azide) | |
| CD29/FNRB/GPIIA/MDF2/MSK12/VLA-BETA/VLAB | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 3688 | |
| Human, Monkey | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| ITGB1 | |
| ITGB1 | |
| A synthetic peptide (conjugated with KLH) corresponding to human ITGB1. | |
| 50 μg | |
| Primary | |
| Extracellular domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only