Learn More
ITGB4 (phospho Y1510), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human ITGB4.
Brand: Abnova PAB29281.100uL
Description
Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Different combinations of alpha and beta polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta 4 subunit, a receptor for the laminins. This subunit tends to associate with alpha 6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq
Specifications
| ITGB4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human ITGB4. | |
| In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| CD104 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3691 | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50∼1:100) Western Blot (1:500∼1:1000) The optimal working dilution should be determined by the end user. | |
| ITGB4 | |
| ITGB4 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y1510 of human ITGB4. | |
| 100 μL | |
| Primary | |
| ITGB4 (phospho Y1510) polyclonal antibody detects endogenous levels of human ITGB4 only when phosphorylated at tyrosine 1510. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only