Learn More
JUP (phospho Y550), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of JUP.
Brand: Abnova PAB7924.100uL
Description
This gene encodes a major cytoplasmic protein which is the only known constituent common to submembranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat. Mutation in this gene has been associated with Naxos disease. Alternative splicing occurs in this gene; however, not all transcripts have been fully described. [provided by RefSeq
Specifications
| JUP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of JUP. | |
| In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
| ARVD12/CTNNG/DP3/DPIII/PDGB/PKGB | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This peptide sequence is highly conserved in rat and mouse g-Catenin. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
| JUP | |
| JUP | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y550 of human JUP. | |
| RUO | |
| 3728 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only