Learn More
KCNC1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KCNC1
Brand: Abnova PAB28508.400uL
Description
The Shaker gene family of Drosophila encodes components of voltage-gated potassium channels and is comprised of four subfamilies. Based on sequence similarity, this gene is similar to one of these subfamilies, namely the Shaw subfamily. The protein encoded by this gene belongs to the delayed rectifier class of channel proteins and is an integral membrane protein that mediates the voltage-dependent potassium ion permeability of excitable membranes. Multiple transcript variants encoding different isoforms have been inferred for this gene based on orthologous loci. [provided by RefSeq
Specifications
| KCNC1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of KCNC1 | |
| In PBS (0.09% sodium azide) | |
| FLJ41162/FLJ42249/FLJ43491/KV3.1/KV4/MGC129855/NGK2 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3746 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| KCNC1 | |
| KCNC1 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 479-508 at C-terminal region at human KCNC1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only