Learn More
KCNK2, Rabbit, DyLight 649, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KCNK2.
Brand: Abnova PAB15123.100uL
Description
This gene encodes one of the members of the two-pore-domain background potassium channel protein family. This type of potassium channel is formed by two homodimers that create a channel that leaks potassium out of the cell to control resting membrane potential. The channel can be opened, however, by certain anesthetics, membrane stretching, intracellular acidosis, and heat. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| KCNK2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of KCNK2. | |
| In 50mM sodium borate | |
| K2p2.1/MGC126742/MGC126744/TPKC1/TREK/TREK-1/TREK1/hTREK-1c/hTREK-1e | |
| Rabbit | |
| 100 μL | |
| Primary | |
| 88% sequence identity with rat protein. TREK 1 Please note that it is essential to run against membrane preps. NOT recommended for immunohistochemistry. | |
| Store at 4°C. Do not freeze. |
| Western Blot | |
| DyLight 649 | |
| Western Blot (2 ug/ mL) The optimal working dilution should be determined by the end user. | |
| KCNK2 | |
| KCNK2 | |
| A synthetic peptide corresponding to amino acids 1-100 of human KCNK2. | |
| RUO | |
| 3776 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only