missing translation for 'onlineSavingsMsg'
Learn More

KDEL, Rabbit, ATTO488, Polyclonal Antibody, Abnova™

Product Code. 16264110
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16264110 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16264110 Supplier Abnova Supplier No. PAB28966.200ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of KDEL.

Specifications

Antigen KDEL
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Atto 488
Description Rabbit polyclonal antibody raised against synthetic peptide of KDEL.
Dilution Immunohistochemistry Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.09% sodium azide)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to KDEL.
Purification Method Affinity purification
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes KDEL proteins, Hsp90b1, Hspa5, PDI and Calr. It may also see PDIA3 and Erp29.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.