missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keratin, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length native Keratin.
Brand: Abnova PAB13720.250uL
This item is not returnable.
View return policy
Specifications
| Keratin | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length native Keratin. | |
| In purified antiserum | |
| Native purified human Keratin. | |
| RUO | |
| Recognizes keratin. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry | |
| Unconjugated | |
| Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 250 μL | |
| Primary | |
| Avian, Human, Mammalia | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction