Learn More
LARP2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LARP2.
Brand: Abnova PAB22975.100uL
Description
This gene encodes a protein containing domains found in the La related protein of Drosophila melanogaster. La motif-containing proteins are thought to be RNA-binding proteins, where the La motif and adjacent amino acids fold into an RNA recognition motif. The La motif is also found in proteins unrelated to the La protein. Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described. Additional splice variation has been identified but the full-length nature of these transcripts has not been determined. [provided by RefSeq
Sequence: KHKWVPLHLDVVRSESQERPGSRNSSRCQPEANKPTHNNRRNDTRSWKRDREKRDDQDDVSSVRSEGGNIRGSFSpecifications
| LARP2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LARP2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp686L13217/MGC117277/MGC75174 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55132 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| LARP2 | |
| LARP2 | |
| Recombinant protein corresponding to amino acids of human LARP2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only