Learn More
LGSN, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LGSN.
Brand: Abnova PAB22768.100uL
Description
This gene encodes a protein with similarity to the GS I members of the glutamine synthetase superfamily. The encoded protein is referred to as a pseudo-glutamine synthetase because it has no glutamine synthesis activity and may function as a chaperone protein. This protein is localized to the lens and may be associated with cataract disease. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: IISFPALTFLNNHDQPFMQELVDGLYHTGANVESFSSSTRPGQMEISFLPEFGISSADNAFTLRTGVKEVARKYNYIASFFIETGFCDSGISpecifications
| LGSN | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LGSN. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| GLULD1/LGS/MGC163238/MGC163240 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51557 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| LGSN | |
| LGSN | |
| Recombinant protein corresponding to amino acids of human LGSN. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only