Learn More
LONP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LONP1.
Brand: Abnova PAB28578.100uL
Description
This gene encodes a mitochondrial matrix protein in the Lon family of ATP-dependent proteases. A similar E. coli protein regulates gene expression by targeting specific regulatory proteins for degradation. This protein binds a specific sequence in the light and heavy chain promoters of the mitochondrial genome which are involved in regulation of DNA replication and transcription. [provided by RefSeq
Sequence: VEEKIKQTHRKYLLQEQLKIIKKELGLEKDDKDAIEEKFRERLKELVVPKHVMDVVDEELSKLGLLDNHSSEFNVTRNYLDWLTSIPWGKYSNENLDLARAQAVLEEDHYGMEDVKKRILEFIAVSQLRGSTQGKILCFYGPSpecifications
| LONP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LONP1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| LON/LONP/LonHS/MGC1498/PIM1/PRSS15/hLON | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9361 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (Human: 1:100-1:250, Mouse/Rat: 1:100-1:500 Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| LONP1 | |
| LONP1 | |
| Recombinant protein corresponding to human LONP1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only