Learn More
LPPR4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LPPR4.
Brand: Abnova PAB10114.100ug
Description
The protein encoded by this gene belongs to the lipid phosphate phosphatase (LPP) family. LPPs catalyze the dephosphorylation of a number of bioactive lipid mediators that regulate a variety of cell functions. This protein is specifically expressed in neurons. It is located in the membranes of outgrowing axons and has been shown to be important for axonal outgrowth during development and regenerative sprouting. [provided by RefSeq
Specifications
| LPPR4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of LPPR4. | |
| In PBS (0.08% sodium azide) | |
| KIAA0455/LPR4/PHP1/PRG-1/PRG1/RP4-788L13.1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western blot (5 to 10 mg/mL) The optimal working dilution should be determined by the end user. | |
| LPPR4 | |
| LPPR4 | |
| A synthetic peptide corresponding to LPPR4. | |
| RUO | |
| 9890 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only