Learn More
LY6G6F, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant LY6G6F.
Brand: Abnova PAB20407.100uL
Description
The human G6f protein is a type I transmembrane protein belonging to the immunoglobin (Ig) superfamily, which is comprised of cell-surface proteins involved in the immune system and cellular recognition (de Vet et al., 2003 [PubMed 12852788]).[supplied by OMIM
Sequence: LLGNYSLWLEGSKEEDAGRYWCAVLGQHHNYQNWRVYDVLVLKGSQLSARAADGSPCNVLLCSVVPSRRMDSVTWQEGKGPVRGRVQSFWGSEAALLLVCPGEGLSEPRSRRPRIIRCLMTHNKGVSFSLAASIDASPALSpecifications
| LY6G6F | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant LY6G6F. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| C6orf21/G6f/LY6G6D/NG32 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 259215 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| LY6G6F | |
| LY6G6F | |
| Recombinant protein corresponding to amino acids of human LY6G6F. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only