Learn More
LYVE1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of LYVE1.
Brand: Abnova PAB3847.400uL
Description
This gene encodes a type I integral membrane glycoprotein. The encoded protein acts as a receptor and binds to both soluble and immobilized hyaluronan. This protein may function in lymphatic hyaluronan transport and have a role in tumor metastasis. [provided by RefSeq
Specifications
| LYVE1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of LYVE1. | |
| In PBS (0.09% sodium azide) | |
| CRSBP-1/HAR/LYVE-1/XLKD1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 10894 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| LYVE1 | |
| LYVE1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human LYVE1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only