Learn More
MAP2K1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAP2K1.
Brand: Abnova PAB18246.100ug
Description
The protein encoded by this gene is a member of the dual specificity protein kinase family, which acts as a mitogen-activated protein (MAP) kinase kinase. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This protein kinase lies upstream of MAP kinases and stimulates the enzymatic activity of MAP kinases upon wide variety of extra- and intracellular signals. As an essential component of MAP kinase signal transduction pathway, this kinase is involved in many cellular processes such as proliferation, differentiation, transcription regulation and development. [provided by RefSeq
Specifications
| MAP2K1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:10000) The optimal working dilution should be determined by the end user. | |
| MAP2K1 | |
| MAP2K1 | |
| A synthetic peptide corresponding to residues surrounding T286 of human MAP2K1. | |
| 100 μg | |
| Primary | |
| This antibody is specific to MAP2K1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of MAP2K1. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| MAPKK1/MEK1/MKK1/PRKMK1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 5604 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only