missing translation for 'onlineSavingsMsg'
Learn More

MAP2K5 Rabbit anti-Chicken, Dog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Product Code. 16060077
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16060077 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16060077 Supplier Abnova Supplier No. PAB12704.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of MAP2K5.

The protein encoded by this gene is a dual specificity protein kinase that belongs to the MAP kinase kinase family. This kinase specifically interacts with and activates MAPK7/ERK5. This kinase itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs). The signal cascade mediated by this kinase is involved in growth factor stimulated cell proliferation and muscle cell differentiation. Four alternatively spliced transcript variants of this gene encoding distinct isoforms have been described. [provided by RefSeq

Specifications

Antigen MAP2K5
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of MAP2K5.
Dilution Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 7.2 (BSA, 10% Proclin300)
Gene MAP2K5
Gene Alias HsT17454/MAPKK5/MEK5/PRKMK5
Gene Symbols MAP2K5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human MAP2K5.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5607
Test Specificity This antibody recognizes ∼49 KDa of human MAP2K5.
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.