Learn More
MAPK11 (phospho T180/T182), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAPK11.
Brand: Abnova PAB15917.100ug
Description
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation, and development. This kinase is most closely related to p38 MAP kinase, both of which can be activated by proinflammatory cytokines and environmental stress. This kinase is activated through its phosphorylation by MAP kinase kinases (MKKs), preferably by MKK6. Transcription factor ATF2/CREB2 has been shown to be a substrate of this kinase. [provided by RefSeq
Specifications
| MAPK11 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAPK11. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| P38B/P38BETA2/PRKM11/SAPK2/SAPK2B/p38-2/p38Beta | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| MAPK11 | |
| MAPK11 | |
| Synthetic phosphopeptide corresponding to residues surrounding T180/T182 of human MAPK11. | |
| RUO | |
| 5600 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only