Learn More
MAPK8IP3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAPK8IP3.
Brand: Abnova PAB26950.100uL
Description
The protein encoded by this gene shares similarity with the product of Drosophila syd gene, required for the functional interaction of kinesin I with axonal cargo. Studies of the similar gene in mouse suggested that this protein may interact with, and regulate the activity of numerous protein kinases of the JNK signaling pathway, and thus function as a scaffold protein in neuronal cells. The C. elegans counterpart of this gene is found to regulate synaptic vesicle transport possibly by integrating JNK signaling and kinesin-1 transport. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq
Specifications
| MAPK8IP3 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| MAPK8IP3 | |
| MAPK8IP3 | |
| A synthetic peptide corresponding to human MAPK8IP3. | |
| 100 μL | |
| Primary | |
| MAPK8IP3 polyclonal antibody detects endogenous levels of MAPK8IP3 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of MAPK8IP3. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| DKFZp762N1113/FLJ00027/JIP3/JSAP1/KIAA1066/SYD2 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 23162 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only