Learn More
MAPKAPK5, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAPKAPK5.
Brand: Abnova PAB2779.400uL
Description
The protein encoded by this gene is a member of the serine/threonine kinase family. In response to cellular stress and proinflammatory cytokines, this kinase is activated through its phosphorylation by MAP kinases including MAPK1/ERK, MAPK14/p38-alpha, and MAPK11/p38-beta. In vitro, this kinase phosphorylates heat shock protein HSP27 at its physiologically relevant sites. Two alternately spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq
Specifications
| MAPKAPK5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MAPKAPK5. | |
| In PBS (0.09% sodium azide) | |
| PRAK | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 8550 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| MAPKAPK5 | |
| MAPKAPK5 | |
| A synthetic peptide (conjugated with KLH) corresponding to residues surrounding T182 of human MAPKAPK5. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only