Learn More
MARCH2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MARCH2.
Brand: Abnova PAB17703.100ug
This item is not returnable.
View return policy
Specifications
| MARCH2. | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MARCH2. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| HSPC240/MARCH-II/RNF172 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total MARCH2 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| MARCH2 | |
| MARCH2 | |
| A synthetic peptide corresponding to internal of human MARCH2. | |
| RUO | |
| 51257 | |
| Human, Mouse, Rat | |
| Liquid |