Learn More
MARCKS (phospho S170), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MARCKS.
Brand: Abnova PAB25909.100ug
Description
The protein encoded by this gene is a substrate for protein kinase C. It is localized to the plasma membrane and is an actin filament crosslinking protein. Phosphorylation by protein kinase C or binding to calcium-calmodulin inhibits its association with actin and with the plasma membrane, leading to its presence in the cytoplasm. The protein is thought to be involved in cell motility, phagocytosis, membrane trafficking and mitogenesis. [provided by RefSeq
Sequence: G-F-Sp-F-KSpecifications
| MARCKS | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| MARCKS | |
| MARCKS | |
| Synthetic phosphopeptide corresponding to residues surrounding S170 of human MARCKS. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of MARCKS. | |
| In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| 80K-L/FLJ14368/FLJ90045/MACS/PKCSL/PRKCSL | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4082 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only