Learn More
MBP Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against MBP.
Brand: Abnova PAB14515.500uL
Description
The protein encoded by the classic MBP gene is a major constituent of the myelin sheath of oligodendrocytes and Schwann cells in the nervous system. However, MBP-related transcripts are also present in the bone marrow and the immune system. These mRNAs arise from the long MBP gene (otherwise called Golli-MBP) that contains 3 additional exons located upstream of the classic MBP exons. Alternative splicing from the Golli and the MBP transcription start sites gives rise to 2 sets of MBP-related transcripts and gene products. The Golli mRNAs contain 3 exons unique to Golli-MBP, spliced in-frame to 1 or more MBP exons. They encode hybrid proteins that have N-terminal Golli aa sequence linked to MBP aa sequence. The second family of transcripts contain only MBP exons and produce the well characterized myelin basic proteins. This complex gene structure is conserved among species suggesting that the MBP transcription unit is an integral part of the Golli transcription unit and that this arrangement is important for the function and/or regulation of these genes. [provided by RefSeq]
Specifications
| MBP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against MBP. | |
| In PBS, pH 7.4 (1% BSA, 0.05% sodium azide) | |
| MGC99675 | |
| Rabbit | |
| 500 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) Immunohistochemistry (Frozen sections) (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| MBP | |
| MBP | |
| Human MBP. | |
| RUO | |
| 4155 | |
| Store at 4°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only