Learn More
MCL1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MCL1.
Brand: Abnova PAB13414.100ug
Description
The protein encoded by this gene belongs to the Bcl-2 family. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. The longer gene product (isoform 1) enhances cell survival by inhibiting apoptosis while the alternatively spliced shorter gene product (isoform 2) promotes apoptosis and is death-inducing. [provided by RefSeq
Specifications
| MCL1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MCL1. | |
| In PBS (0.02% sodium azide) | |
| BCL2L3/EAT/MCL1L/MCL1S/MGC104264/MGC1839/TM | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| MCL1 | |
| MCL1 | |
| A synthetic peptide corresponding to C-terminus 16 amino acids of human MCL1. | |
| RUO | |
| 4170 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only