missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2 (phospho S454), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MEF2.
Brand: Abnova PAB15920.100ug
This item is not returnable.
View return policy
Specifications
| MEF2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of MEF2. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| Synthetic phosphopeptide corresponding to residues surrounding S454 of human MEF2. | |
| RUO | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction