Learn More
METTL7A Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of METTL7A.
Brand: Abnova PAB16591.100ug
This item is not returnable.
View return policy
Specifications
| METTL7A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of METTL7A. | |
| In PBS (0.02% sodium azide) | |
| AAM-B/DKFZp586A0522 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| METTL7A | |
| METTL7A | |
| A synthetic peptide corresponding to internal region 13 amino acids of human METTL7A. | |
| RUO | |
| 25840 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |