missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFAP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant MFAP1.
Brand: Abnova PAB23931.100uL
This item is not returnable.
View return policy
Description
Sequence: KELEENKRSLAALDALNTDDENDEEEYEAWKVRELKRIKRDREDREALEKEKAEIERMRNLTEEERRAELRANGKVITNKAVKGKYKFLQKYYHRGAFFMDEDEEVYKRDFSAPTLEDHFNKTILPKVMQVKNFGRSGTekniske data
| MFAP1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant MFAP1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| MFAP1 | |
| Recombinant protein corresponding to amino acids of human MFAP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:2500-1:5000) The optimal working dilution should be determined by the end user. | |
| MFAP1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 4236 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion