Learn More
MGAT1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MGAT1.
Brand: Abnova PAB4304.400uL
Description
There are believed to be over 100 different glycosyltransferases involved in the synthesis of protein-bound and lipid-bound oligosaccharides. UDP-N-acetylglucosamine:alpha-3-D-mannoside beta-1,2-N-acetylglucosaminyltransferase I is a medial-Golgi enzyme essential for the synthesis of hybrid and complex N-glycans. The protein, encoded by a single exon, shows typical features of a type II transmembrane protein. The protein is believed to be essential for normal embryogenesis. Several variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
| MGAT1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MGAT1. | |
| In PBS (0.09% sodium azide) | |
| GLCNAC-TI/GLCT1/GLYT1/GNT-1/GNT-I/MGAT | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 4245 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| MGAT1 | |
| MGAT1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human MGAT1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only