Learn More
MINK1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MINK1.
Brand: Abnova PAB16441.50ug
Beskrivning
This gene encodes a serine/threonine kinase belonging to the germinal center kinase (GCK) family. The protein is structurally similar to the kinases that are related to NIK and may belong to a distinct subfamily of NIK-related kinases within the GCK family. Studies of the mouse homolog indicate an up-regulation of expression in the course of postnatal mouse cerebral development and activation of the cJun N-terminal kinase (JNK) and the p38 pathways. Alternative splicing occurs at this locus and four transcript variants encoding distinct isoforms have been identified. [provided by RefSeq
Specifikationer
| MINK1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MINK1. | |
| In PBS (0.09% sodium azide) | |
| B55/MAP4K6/MGC21111/MINK/YSK2/ZC3/hMINK/hMINKbeta | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 50488 | |
| Equine, Hamster, Human, Monkey, Mouse, Pig, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| MINK1 | |
| MINK1 | |
| A synthetic peptide (conjugated with KLH) corresponding to human MINK1. | |
| 50 μg | |
| Primary | |
| Internal domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only