Learn More
MMP16 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MMP16.
Brand: Abnova PAB4784.400uL
Description
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene produces at least two transcripts, one which encodes a membrane-bound form and another a soluble form of the protein. Both forms of the protein activate MMP2 by cleavage. This gene was once referred to as MT-MMP2, but was renamed as MT-MMP3 or MMP16. [provided by RefSeq
Specifications
| MMP16 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MMP16. | |
| In PBS (0.09% sodium azide) | |
| C8orf57/MMP-X2/MT-MMP2/MT-MMP3/MT3-MMP | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 4325 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| MMP16 | |
| MMP16 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human MMP16. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only