missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkey Rhesus rotavirus Outer capsid protein VP4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Monkey Rhesus rotavirus Outer capsid protein VP4.
Brand: Abnova PAB16097.100ug
This item is not returnable.
View return policy
Specifications
| Monkey Rhesus rotavirus Outer capsid protein VP4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Monkey Rhesus rotavirus Outer capsid protein VP4. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| A synthetic peptide corresponding to N-terminus of Monkey Rhesus rotavirus Outer capsid protein VP4. | |
| RUO | |
| Virus | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction