missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG F(ab')2 fragment,Polyclonal Secondary Antibody, Abnova™
Brand: Abnova PAB9363.2mg
This item is not returnable.
View return policy
Specifications
| Rabbit anti-Mouse IgG F(ab')2 fragment | |
| Polyclonal | |
| Rabbit anti-mouse IgG recognizes mouse IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Mouse IgG F(ab')2 fragment | |
| 2 mg | |
| Mouse | |
| Liquid |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction