Learn More
Rabbit anti-Mouse IgG F(ab')2 Secondary Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-72782UV
Product Code. 30231664
  • € 342.00 / 0.50mL
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG F(ab')2
Polyclonal
50 mM Sodium Borate
Mouse IgG F(ab')2 fragment
0.5 mL
Secondary
Store at 4°C in the dark.
Western Blot, ELISA, Immunohistochemistry
DyLight 350
Rabbit
Multi-step