Learn More
Rabbit anti-Mouse IgG F(ab')2 Secondary Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72782H
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| PBS | |
| Mouse IgG F(ab')2 fragment | |
| 0.5 mL | |
| Secondary | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry | |
| HRP | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Mouse | |
| IgG |