missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG1 Secondary Antibody (20) [DyLight 594], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43768DL594
This item is not returnable.
View return policy
Description
IgG1 Monoclonal antibody specifically detects IgG1 in Mouse samples. It is validated for ELISA
Specifications
| IgG1 | |
| Monoclonal | |
| DyLight 594 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Mouse | |
| Purified |
| ELISA | |
| 20 | |
| ELISA | |
| IgG1, Igh-4, RGD1359539, VH7183 | |
| Recombinant Mouse IgG1 Fc Protein | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion