missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Mouse IgG2b Heavy Chain Secondary Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-72795PCP
This item is not returnable.
View return policy
Description
IgG2b Heavy Chain Polyclonal antibody specifically detects IgG2b Heavy Chain in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen), Fluorophore-linked immunosorbent assay
Specifications
| IgG2b Heavy Chain | |
| Polyclonal | |
| PBS | |
| Mouse IgG2b heavy chain | |
| 0.5 mL | |
| Secondary | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen), Fluorophore-Linked Immunosorbent Assay | |
| PerCP | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction