missing translation for 'onlineSavingsMsg'
Learn More

Mrc1, Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16192060
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16192060 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16192060 Supplier Abnova Supplier No. PAB15587.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against partial recombinant Mrc1.

Specifications

Antigen Mrc1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Mrc1.
Dilution Western Blot (1:3000) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.05% sodium azide)
Gene mrc1
Gene Symbols mrc1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-317 Schizosaccharomyces pombe Mrc1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2543240
Target Species S. pombe
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.