Learn More
MRPL39, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MRPL39.
Brand: Abnova PAB17770.100ug
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Two transcript variants encoding distinct isoforms have been described. A pseudogene corresponding to this gene is found on chromosome 5q. [provided by RefSeq
Specifications
| MRPL39 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MRPL39. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| C21orf92/FLJ20451/L39mt/MGC104174/MGC3400/MRP-L5/MRPL5/MSTP003/PRED22/PRED66/RPML5 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total MRPL39 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| MRPL39 | |
| MRPL39 | |
| A synthetic peptide corresponding to internal of human MRPL39. | |
| RUO | |
| 54148 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only