Learn More
MRPL47 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MRPL47.
Brand: Abnova PAB17565.100ug
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. This gene is immediately adjacent to the gene for BAF complex 53kDa subunit protein a (BAF53a), in a tail-to-tail orientation. Two transcript variants encoding different protein isoforms have been identified. [provided by RefSeq
Specifications
| MRPL47 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MRPL47. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| CGI-204/MGC45403/NCM1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total MRPL47 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
| MRPL47 | |
| MRPL47 | |
| A synthetic peptide corresponding to internal of human MRPL47. | |
| RUO | |
| 57129 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only