Learn More
MS4A1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MS4A1.
Brand: Abnova PAB12666.100ug
Description
This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. This gene encodes a B-lymphocyte surface molecule which plays a role in the development and differentiation of B-cells into plasma cells. This family member is localized to 11q12, among a cluster of family members. Alternative splicing of this gene results in two transcript variants which encode the same protein. [provided by RefSeq
Specifications
| MS4A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MS4A1. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| B1/Bp35/CD20/LEU-16/MGC3969/MS4A2/S7 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes ∼34 KDa of human MS4A1. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| MS4A1 | |
| MS4A1 | |
| A synthetic peptide corresponding to C-terminus of human MS4A1. | |
| RUO | |
| 931 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only