Learn More
MUC1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MUC1.
Brand: Abnova PAB18319.100ug
Description
This gene is a member of the mucin family and encodes a membrane bound, glycosylated phosphoprotein. The protein is anchored to the apical surface of many epithelia by a transmembrane domain, with the degree of glycosylation varying with cell type. It also includes a 20 aa variable number tandem repeat (VNTR) domain, with the number of repeats varying from 20 to 120 in different individuals. The protein serves a protective function by binding to pathogens and also functions in a cell signaling capacity. Overexpression, aberrant intracellular localization, and changes in glycosylation of this protein have been associated with carcinomas. Multiple alternatively spliced transcript variants that encode different isoforms of this gene have been reported, but the full-length nature of only some has been determined. [provided by RefSeq
Specifications
| MUC1 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
| MUC1 | |
| MUC1 | |
| A synthetic peptide corresponding to human MUC1. | |
| 100 μg | |
| Primary | |
| This antibody is specific to MUC1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of MUC1. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| CD227/EMA/H23AG/MAM6/PEM/PEMT/PUM | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 4582 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only