missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC12 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MUC12.
Brand: Abnova PAB12553.100ug
This item is not returnable.
View return policy
Description
Sequence: WEDQNLRESRFGLENSpecifications
| MUC12 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MUC12. | |
| In PBS (0.02% sodium azide) | |
| MUC11 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Reacts with residues 541-555 [WEDQNLRESRFGLEN] of the human MUC12 protein. | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| ELISA (1:500-1:1000) Immunofluorescence (1:500-1:1000) Immunohistochemistry (Frozen sections) (1:500-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1:1000) Immunoprecipitation (1:500-1:1000) Western Blot | |
| MUC12 | |
| MUC12 | |
| A synthetic peptide corresponding to amino acids 541-555 of human MUC12. | |
| RUO | |
| 10071 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction