missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYD88 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MYD88.
Brand: Abnova PAB13080.100ug
This item is not returnable.
View return policy
Specifications
| MYD88 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MYD88. | |
| In PBS (0.02% sodium azide) | |
| MYD88D | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Differ from those of mouse by two amino acids. an approximately 35 KDa band can be detected. It is human and mouse reactive. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| MYD88 | |
| MYD88 | |
| A synthetic peptide corresponding to amino acids 233-248 of human MYD88. | |
| RUO | |
| 4615 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction