missing translation for 'onlineSavingsMsg'
Learn More

MYST4, Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16213340
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16213340 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16213340 Supplier Abnova Supplier No. PAB13579.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of MYST4.

Specifications

Antigen MYST4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of MYST4.
Dilution Western Blot (1:7500) Immunohistochemistry (1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4
Gene MYST4
Gene Symbols MYST4
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to bovine MYST4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 535403
Target Species Bovine
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 125μL deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.