missing translation for 'onlineSavingsMsg'
Learn More

NBN Rabbit anti-Human, Mouse, Primates, Polyclonal Antibody, Abnova™

Product Code. 16089776
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16089776 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16089776 Supplier Abnova Supplier No. PAB12377.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against partial recombinant NBN.

Mutations in this gene are associated with Nijmegen breakage syndrome, an autosomal recessive chromosomal instability syndrome characterized by microcephaly, growth retardation, immunodeficiency, and cancer predisposition. The encoded protein is a member of the MRE11/RAD50 double-strand break repair complex which consists of 5 proteins. This gene product is thought to be involved in DNA double-strand break repair and DNA damage-induced checkpoint activation. [provided by RefSeq

Specifications

Antigen NBN
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant NBN.
Dilution Immunocytochemistry (1:50-1:200) Immunofluorescence (1:200) Immunoprecipitation (3 ug/mL) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.09% sodium azide)
Gene NBN
Gene Alias AT-V1/AT-V2/ATV/FLJ10155/MGC87362/NBS/NBS1/P95
Gene Symbols NBN
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-705 of human NBN.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4683
Test Specificity This antibody is specific to human NBS1.
Target Species Human, Mouse, Primate
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.