Learn More
NEFH Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NEFH.
Brand: Abnova PAB25167.100uL
Description
Neurofilaments are type IV intermediate filament heteropolymers composed of light, medium, and heavy chains. Neurofilaments comprise the axoskeleton and functionally maintain neuronal caliber. They may also play a role in intracellular transport to axons and dendrites. This gene encodes the heavy neurofilament protein. This protein is commonly used as a biomarker of neuronal damage and susceptibility to amyotrophic lateral sclerosis (ALS) has been associated with mutations in this gene. [provided by RefSeq
Specifications
| NEFH | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| NEFH | |
| NEFH | |
| A synthetic peptide corresponding to NEFH. | |
| 100 μL | |
| Primary | |
| NEFH polyclonal antibody detects endogenous levels of NEFH protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of NEFH. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| NFH | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 4744 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only