missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEURL2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NEURL2.
Brand: Abnova PAB17799.100ug
This item is not returnable.
View return policy
Specifications
| NEURL2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NEURL2. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| C20orf163/MGC125934/MGC125935/OZZ/Ozz-E3 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total NEURL2 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| NEURL2 | |
| NEURL2 | |
| A synthetic peptide corresponding to internal of human NEURL2. | |
| RUO | |
| 140825 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction