missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFKB1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NFKB1.
Brand: Abnova PAB16888.100ug
This item is not returnable.
View return policy
Description
Sequence: QLRRKSpDLETSEPSpecifications
| NFKB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NFKB1. | |
| Lyophilized from 10mM PBS | |
| A synthetic peptide (conjugated with KLH) corresponding to NFKB1. | |
| RUO | |
| 4970 | |
| Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA | |
| Unconjugated | |
| ELISA (1:100000-1:400000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction