missing translation for 'onlineSavingsMsg'
Learn More

NHLRC3, Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16183710
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16183710 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16183710 Supplier Abnova Supplier No. PAB20670.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against recombinant NHLRC3.

Sequence: ILWLHGENGTGPAKFNIPHSVTLDSAGRVWVADRGNKRIQVFDKDTGEWLGAWNNCFTEEGPSSVRFTPDGKYLIVAQLNLSRLSVVAAPPVGSIGECSVISTIQLADQ

Specifications

Antigen NHLRC3
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant NHLRC3.
Dilution Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene NHLRC3
Gene Alias DKFZp313M1221/DKFZp686E1140
Gene Symbols NHLRC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human NHLRC3.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387921
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.